KEY465197566 Key Organics / BIONET

7-chloro-N-(4-methylphenyl)-3,4-dihydro-2H-1,4-benzothiazine-4-carboxamide

  • MDL number MFCD00138834
  • Cas Number : 338777-89-2
  • Molecular Weight : 318.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81194

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6B-075_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart