KEY465197575 Key Organics / BIONET

3-{[(4-chlorophenyl)methyl]sulfanyl}-5-methyl-1,2-thiazole-4-carbonitrile

  • MDL number MFCD00138524
  • Cas Number : 338778-09-9
  • Molecular Weight : 280.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81218

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6C-008_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart