KEY465197586 Key Organics / BIONET

methyl 2-[({4-cyano-5-[(E)-2-(dimethylamino)ethenyl]-1,2-thiazol-3-yl}sulfanyl)methyl]benzoate

  • MDL number MFCD00138526
  • Cas Number : 338778-45-3
  • Molecular Weight : 359.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81254

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6C-054_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart