KEY465197596 Key Organics / BIONET

3-{[(2-methyl-1,3-thiazol-4-yl)methyl]sulfanyl}-5-[(E)-2-(phenylamino)ethenyl]-1,2-thiazole-4-carbonitrile

  • MDL number MFCD00138547
  • Cas Number : 338778-89-5
  • Molecular Weight : 370.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81279

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6C-083_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart