KEY465197626 Key Organics / BIONET

methyl (2E)-2-(phenylformamido)-3-{[3-(trifluoromethyl)phenyl]amino}prop-2-enoate

  • MDL number MFCD00172774
  • Cas Number : 338785-15-2
  • Molecular Weight : 364.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl (2E)-2-(benzoylamino)-3-[3-(trifluoromethyl)anilino]-2-propenoate
Inchi Code 1S/C18H15F3N2O3/c1-26-17(25)15(23-16(24)12-6-3-2-4-7-12)11-22-14-9-5-8-13(10-14)18(19,20)21/h2-11,22H,1H3,(H,23,24)/b15-11+
Inchi Key XRGPASOWJNAVKA-RVDMUPIBSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81516

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/6E-907_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart