KEY465197629 Key Organics / BIONET

methyl (3E)-2-[(2,4-dichlorophenyl)formamido]-3-(methoxyimino)propanoate

  • MDL number MFCD00141032
  • Cas Number : 318517-58-7
  • Molecular Weight : 319.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl (3E)-2-[(2,4-dichlorobenzoyl)amino]-3-(methoxyimino)propanoate
Inchi Code 1S/C12H12Cl2N2O4/c1-19-12(18)10(6-15-20-2)16-11(17)8-4-3-7(13)5-9(8)14/h3-6,15H,1-2H3,(H,16,17)
Inchi Key HQYSQQHMRNKMJV-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81530

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/6E-933_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart