KEY465197634 Key Organics / BIONET

4-[5-(2,4-dichlorophenyl)furan-2-yl]-1,3-thiazol-2-amine

  • MDL number MFCD00202423
  • Cas Number : 338785-87-8
  • Molecular Weight : 311.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81564

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6F-028_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart