KEY465197657 Key Organics / BIONET

1,3-diethyl 2-{[(4-benzoyl-1,5-dimethyl-1H-pyrrol-3-yl)amino]methylidene}propanedioate

  • MDL number MFCD00171821
  • Cas Number : 131169-02-3
  • Molecular Weight : 384.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81694

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6F-950_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart