KEY465197662 Key Organics / BIONET

3-(4-hydroxyphenoxy)-5-(trifluoromethyl)pyridine-2-carbonitrile

  • MDL number MFCD00664131
  • Cas Number : 338791-69-8
  • Molecular Weight : 280.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81726

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6G-031_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart