KEY465197670 Key Organics / BIONET

ethyl 2,2-difluoro-2-[3-(trifluoromethyl)phenoxy]acetate

  • MDL number MFCD00173140
  • Cas Number : 338791-94-9
  • Molecular Weight : 284.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81769

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6G-324S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart