KEY465197675 Key Organics / BIONET

1-(4-bromophenyl)-4-methyl-4-nitropentan-1-one

  • MDL number MFCD01936384
  • Cas Number : 400085-27-0
  • Molecular Weight : 300.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81806

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6G-391S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart