KEY465197676 Key Organics / BIONET

methyl 2,2-difluoro-2-(4-fluorophenoxy)acetate

  • MDL number MFCD00173165
  • Cas Number : 338792-43-1
  • Molecular Weight : 220.147

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Store at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81827

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404,P411
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6G-416S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart