KEY465197678 Key Organics / BIONET

propan-2-yl 2-[4-(2-fluorophenyl)piperazin-1-yl]pyridine-3-carboxylate

  • MDL number MFCD00244889
  • Cas Number : 338792-74-8
  • Molecular Weight : 343.402

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81866

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6G-466S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart