KEY465197699 Key Organics / BIONET

5-(2-chloro-6-fluorophenyl)-3-hydroxy-2-(1,2,3,4-tetrahydroisoquinolin-1-yl)cyclohex-2-en-1-one

  • MDL number MFCD00215034
  • Cas Number : 338793-14-9
  • Molecular Weight : 371.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81935

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6H-028_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart