KEY465197703 Key Organics / BIONET

ethyl 2-sulfanylidene-5-({[3-(trifluoromethyl)phenyl]methyl}sulfanyl)-2H-1,3-dithiole-4-carboxylate

  • MDL number MFCD00173478
  • Cas Number : 338793-29-6
  • Molecular Weight : 396.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81954

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6H-050_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart