KEY465197709 Key Organics / BIONET

6-(thiophen-2-yl)-[1,2,4]triazolo[1,5-a]pyrimidine-2,7-diamine

  • MDL number MFCD00232516
  • Cas Number : 338793-64-9
  • Molecular Weight : 232.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81975

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6H-077_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart