KEY465197725 Key Organics / BIONET

2-amino-4-(2-chlorophenyl)-6-(2-methylpropyl)pyridine-3-carbonitrile

  • MDL number MFCD00244166
  • Cas Number : 337474-61-0
  • Molecular Weight : 285.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82022

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6H-357S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart