KEY465197729 Key Organics / BIONET

2-amino-4-(4-chlorophenyl)-6-ethyl-5-methylpyridine-3-carbonitrile

  • MDL number MFCD00244171
  • Cas Number : 338794-05-1
  • Molecular Weight : 271.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82031

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6H-370S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart