KEY465197736 Key Organics / BIONET

2-amino-6-(4-chlorophenyl)-4-phenylpyridine-3-carbonitrile

  • MDL number MFCD00244181
  • Cas Number : 75356-07-9
  • Molecular Weight : 305.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82049

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6H-393S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart