KEY465197757 Key Organics / BIONET

ethyl 2-(N-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl](methyl)amino}carbamimidoyl)acetate

  • MDL number MFCD00244787
  • Cas Number : 338794-86-8
  • Molecular Weight : 338.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82126

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6H-923_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart