KEY465197783 Key Organics / BIONET

ethyl 4,5,8-trichloroquinoline-3-carboxylate

  • MDL number MFCD00173386
  • Cas Number : 338795-11-2
  • Molecular Weight : 304.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Hygroscopic
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82181

Safety Information

signalword Warning
pcodes P232, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6J-025_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart