KEY465197785 Key Organics / BIONET

ethyl 8-chloro-4-hydroxy-6-methylquinoline-3-carboxylate

  • MDL number MFCD00173392
  • Cas Number : 338795-13-4
  • Molecular Weight : 265.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82187

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/6J-031_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart