KEY465197808 Key Organics / BIONET

1-methyl-2-oxo-5-[3-(trifluoromethyl)phenyl]-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD00232375
  • Cas Number : 338964-42-4
  • Molecular Weight : 278.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82278

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6J-516S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart