KEY465197816 Key Organics / BIONET

1-[(2,6-dichlorophenyl)methyl]-5-(4-methylphenyl)-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD00232521
  • Cas Number : 338964-95-7
  • Molecular Weight : 369.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82322

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6J-588S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart