KEY465197819 Key Organics / BIONET

3-(pyridin-3-yl)-6-(trifluoromethyl)thieno[3,2-b]pyridine-2-carbohydrazide

  • MDL number MFCD01315035
  • Cas Number : 672951-23-4
  • Molecular Weight : 338.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82337

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6K-001_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart