KEY465197850 Key Organics / BIONET

ethyl 5-[(3-chlorophenyl)sulfanyl]-3-(4-methylphenyl)-1,2,4-triazine-6-carboxylate

  • MDL number MFCD00664970
  • Cas Number : 338965-79-0
  • Molecular Weight : 385.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82472

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6K-529S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart