KEY465197855 Key Organics / BIONET

1-[1-methyl-3-phenyl-5-(phenylsulfanyl)-1H-pyrazol-4-yl]hept-2-yn-1-ol

  • MDL number MFCD00664982
  • Cas Number : 318949-14-3
  • Molecular Weight : 376.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82504

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6K-581S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart