KEY465197859 Key Organics / BIONET

N-(3-acetylphenyl)-2-chlorobenzene-1-sulfonamide

  • MDL number MFCD00664893
  • Cas Number : 338966-22-6
  • Molecular Weight : 309.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82535

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6K-652S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart