KEY465197887 Key Organics / BIONET

(2E)-2-[5-(4-chlorobenzoyl)thiophen-2-yl]-3-(3-methylthiophen-2-yl)prop-2-enenitrile

  • MDL number MFCD00664576
  • Cas Number : 338966-69-1
  • Molecular Weight : 369.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82611

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6L-025_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart