KEY465197908 Key Organics / BIONET

1-(4-chloro-3-nitrophenyl)-4-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]quinoxaline

  • MDL number MFCD00974713
  • Cas Number : 343372-41-8
  • Molecular Weight : 393.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82820

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6M-013_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart