KEY465197921 Key Organics / BIONET

1-[(4-methylphenyl)amino]-3-phenyl-2,5-dihydro-1H-pyrrole-2,5-dione

  • MDL number MFCD00974730
  • Cas Number : 271254-70-7
  • Molecular Weight : 278.311

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82844

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6M-046_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart