KEY465197922 Key Organics / BIONET

2-(2-nitroethyl)-N-[3-(trifluoromethyl)phenyl]benzamide

  • MDL number MFCD00974349
  • Cas Number : 343372-64-5
  • Molecular Weight : 338.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82848

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6M-050_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart