KEY465197923 Key Organics / BIONET

methyl 4-[2-(2-nitroethyl)benzamido]benzoate

  • MDL number MFCD00974350
  • Cas Number : 343372-65-6
  • Molecular Weight : 328.324

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82849

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6M-051_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart