KEY465197926 Key Organics / BIONET

4-chloro-N-{[3-(2-chlorophenyl)-1,2-oxazol-5-yl]methyl}benzene-1-sulfonamide

  • MDL number MFCD01315715
  • Cas Number : 343372-71-4
  • Molecular Weight : 383.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82882

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6M-349S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart