KEY465197931 Key Organics / BIONET

2-chloro-N-{[3-(4-chlorophenyl)-1,2-oxazol-5-yl]methyl}-6-methylbenzene-1-sulfonamide

  • MDL number MFCD01315727
  • Cas Number : 343372-83-8
  • Molecular Weight : 397.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82906

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6M-373S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart