KEY465197933 Key Organics / BIONET

methyl 2-(4-methylbenzenesulfonamido)-3-phenylpropanoate

  • MDL number MFCD00563406
  • Cas Number : 51220-86-1
  • Molecular Weight : 333.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82920

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6M-538S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart