KEY465197938 Key Organics / BIONET

N-(2-{[(4-chlorophenyl)methyl]sulfanyl}-2-methylpropyl)-4-fluorobenzene-1-sulfonamide

  • MDL number MFCD00975600
  • Cas Number : 343373-00-2
  • Molecular Weight : 387.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82963

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6M-728_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart