KEY465197941 Key Organics / BIONET

(3E)-4-[(3,5-dichlorophenyl)amino]-3-phenylbut-3-en-2-one

  • MDL number MFCD00975145
  • Cas Number : 343373-03-5
  • Molecular Weight : 306.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82966

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6M-734_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart