KEY465197958 Key Organics / BIONET

(Z)-N'-hydroxy-N-(4-methoxyphenyl)-3-nitrobenzene-1-carboximidamide

  • MDL number MFCD27924219
  • Cas Number : 299202-70-3
  • Molecular Weight : 287.275

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170 - 171
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/402892

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6N-048_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart