KEY465197961 Key Organics / BIONET

1-benzoyl-3-(quinolin-8-yl)thiourea

  • MDL number MFCD01568682
  • Cas Number : 130818-06-3
  • Molecular Weight : 307.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83039

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6N-056_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart