KEY465197964 Key Organics / BIONET

(isoquinolin-5-yl)thiourea

  • MDL number MFCD01568685
  • Cas Number : 72677-72-6
  • Molecular Weight : 203.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83044

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6N-064_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart