KEY465197979 Key Organics / BIONET

1,1-dioxo-2H,3H,4H-1lambda6-thieno[2,3-b]thiopyran-4-yl 2,4-dichlorobenzoate

  • MDL number MFCD01568370
  • Cas Number : 343373-81-9
  • Molecular Weight : 377.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83106

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6N-359S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart