KEY465198002 Key Organics / BIONET

(E)-1-[(2E)-3-(2-chloro-6-fluorophenyl)-1-(4-fluorophenyl)prop-2-en-1-ylidene]-2-(4-fluorophenyl)hydrazine

  • MDL number MFCD01316710
  • Cas Number : 343374-70-9
  • Molecular Weight : 386.80

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83246

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6N-700_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart