KEY465198003 Key Organics / BIONET

1,3-bis(4-fluorophenyl)-5-(4-methylphenyl)-4,5-dihydro-1H-pyrazole

  • MDL number MFCD01443654
  • Cas Number : 343374-71-0
  • Molecular Weight : 348.397

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83247

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6N-701_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart