KEY465198026 Key Organics / BIONET

ethyl 3-[(3-chlorophenyl)methoxy]pyrido[1,2-a]indole-10-carboxylate

  • MDL number MFCD01814932
  • Cas Number : 672951-93-8
  • Molecular Weight : 379.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83318

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6N-825_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart