KEY465198036 Key Organics / BIONET

(E)-N'-[(1Z)-2-amino-1,2-dicyanoeth-1-en-1-yl]-N-(2-ethoxyphenyl)methanimidamide

  • MDL number MFCD01871566
  • Cas Number : 672952-01-1
  • Molecular Weight : 255.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83339

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6P-025_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart