KEY465198050 Key Organics / BIONET

5-hydroxy-1,3-oxathiolane-2-carboxylic acid

  • MDL number MFCD01871573
  • Cas Number : 672952-08-8
  • Molecular Weight : 150.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83370

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6P-110_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart