KEY465198053 Key Organics / BIONET

2-methoxy-N'-[3-(1H-pyrrol-1-yl)thiophene-2-carbonyl]benzohydrazide

  • MDL number MFCD02186686
  • Cas Number : 672952-11-3
  • Molecular Weight : 341.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83381

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6P-312S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart