KEY465198075 Key Organics / BIONET

4-fluoro-N'-[2-(1H-pyrrol-1-yl)benzoyl]benzohydrazide

  • MDL number MFCD02102709
  • Cas Number : 478063-01-3
  • Molecular Weight : 323.327

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83418

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6P-358S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart