KEY465198080 Key Organics / BIONET

N'-(4-methoxybenzenesulfonyl)-2-(1H-pyrrol-1-yl)benzohydrazide

  • MDL number MFCD02102713
  • Cas Number : 478063-15-9
  • Molecular Weight : 371.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83437

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6P-402S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart